missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Dichlorophosphate, TRC

Product Code. 30417712
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
30417712 25 g 25g
30410993 100 g 100g
2 options
This item is not returnable. View return policy

Product Code. 30417712

Brand: Toronto Research Chemicals TRCP31995025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenyl Dichlorophosphate
CAS 770-12-7
Molecular Formula C6H5Cl2O2P
Quantity 25 g
Synonym Phosphorodichloridic Acid Phenyl Ester, Phenyl Phosphorodichloridate, Dichlorophenoxyphosphine Oxide, NSC 44412, Phenoxydichlorophosphine Oxide, Phenoxyphosphoryl Dichloride, Phenyl Dichlorophosphinate, Phenylphosphoric Dichloride
InChI Formula InChI=1S/C6H5Cl2O2P/c7-11(8,9)10-6-4-2-1-3-5-6/h1-5H
SMILES ClP(=O)(Cl)Oc1ccccc1
IUPAC Name dichlorophosphoryloxybenzene
Molecular Weight (g/mol) 210.98
Formula Weight 209.9404
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.