missing translation for 'onlineSavingsMsg'
Learn More

Phenyl propargyl ether, 98+%

Product Code. 11493817
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493817 5 g 5g
11483817 1 g 1g
11403827 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493817 Supplier Thermo Scientific Alfa Aesar Supplier No. A18272.06
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13610-02-1 | C9H8O | 132.16 g/mol

Phenyl propargyl ether is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl propargyl ether is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water (1.0 g/L at 25°C).

Notes
Air and light sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13610-02-1
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.16
MDL Number MFCD00048107
InChI Key AIQRJSXKXVZCJO-UHFFFAOYSA-N
Synonym phenyl propargyl ether, prop-2-ynyloxy benzene, prop-2-yn-1-yloxy benzene, phenyl 2-propynyl ether, benzene, 2-propynyloxy, 3-phenoxy-1-propyne, 2-propynyloxy benzene, phenylpropargyl ether, prop-2-ynyloxybenzene, ether, phenyl 2-propynyl
PubChem CID 83613
IUPAC Name prop-2-ynoxybenzene
SMILES C#CCOC1=CC=CC=C1

Specifications

Density 1.03
Boiling Point 89°C to 90°C (14 mmHg)
Flash Point 76°C (169°F)
Odor Pungent
Linear Formula C6H5OCH2C≡CH
Refractive Index 1.533
Quantity 5 g
Sensitivity Air and light sensitive
Solubility Information Slightly soluble in water (1.0g/L at 25°C).
Formula Weight 132.16
Percent Purity ≥98%
Chemical Name or Material Phenyl propargyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.