missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Phenyl Sepharose™ 6 Fast Flow Chromatography Media

Product Code. 10505210 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
200 mL
Product Type:
Phenyl Sepharose 6 Fast Flow Media (high sub)
Phenyl Sepharose 6 Fast Flow Media (low sub)
Unit Size:
200mL
25mL
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Type unitSize
10505210 200 mL Phenyl Sepharose 6 Fast Flow Media (low sub) Each
10343340 200 mL Phenyl Sepharose 6 Fast Flow Media (high sub) 200mL
10231400 25 mL Phenyl Sepharose 6 Fast Flow Media (low sub) Each
10348259 25 mL Phenyl Sepharose 6 Fast Flow Media (high sub) 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10505210 Supplier Cytiva Supplier No. 17096505

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for capture and intermediate purification requiring low to medium hydrophobicity

  • Base matrix derivatized via uncharged, chemically-stable ether linkages
  • Two levels of substitution help find optimal selectivity and binding capacity for given application
  • Hydrophilic nature of the base matrix ensures low levels of non-specific binding
  • Hydrophobic interaction chromatography (HIC) media composed of cross-linked 6% agarose beads modified with standard aromatic phenyl groups via uncharged, chemically-stable ether linkages
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
  • Degree of substitution for low sub HIC media is ∽25μmol phenyl/mL gel
  • Degree of substitution for high sub HIC media is ∽40μmol phenyl/mL gel

TRUSTED_SUSTAINABILITY

Specifications

Quantity 200 mL
For Use With (Application) For Capture and Intermediate PurifiCatIon Requiring Low to Medium Hydrophobicity
Product Type Phenyl Sepharose 6 Fast Flow Media (low sub)
Flow Rate 200 to 400 cm/hr
Matrix 6% Cross-Linked Agarose
pH Range 3 to 13
Ligand Type Phenyl
compliance-icons
Product Identifier
  • Phenyl Sepharose 6 Fast Flow (low sub), 200 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.