missing translation for 'onlineSavingsMsg'
Learn More

Phenyl sulfide, 99%

Product Code. 10193130 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10193130 25 mL Glass bottle 25mL
10591951 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10193130 Supplier Thermo Scientific Acros Supplier No. 130950250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 139-66-2 | C12H10S | 186.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-66-2
Molecular Formula C12H10S
Molecular Weight (g/mol) 186.27
MDL Number MFCD00003064
InChI Key LTYMSROWYAPPGB-UHFFFAOYSA-N
Synonym diphenyl sulfide, phenyl sulfide, phenylthiobenzene, diphenyl sulphide, diphenyl thioether, benzene, 1,1'-thiobis, diphenylmercaptan, diphenylthiamethane, diphenyl monosulfide, sulfide, diphenyl
PubChem CID 8766
ChEBI CHEBI:38959
IUPAC Name phenylsulfanylbenzene
SMILES S(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point -40°C
Color Colorless to Yellow
Density 1.1100g/mL
Boiling Point 296°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (C6H5)2S
Refractive Index 1.6317 to 1.6337
Quantity 25 mL
Beilstein 06, 299
Fieser 06,241
Merck Index 15, 3371
Specific Gravity 1.11
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in hot alcohol,miscible with benzene,ether and carbon disulfide
Formula Weight 186.27
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Phenyl sulfide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Phenyl sulfide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.