missing translation for 'onlineSavingsMsg'
Learn More

Phenyl sulfoxide, 97%

Product Code. 10616722 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10616722 100 g Glass bottle 100g
10407720 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10616722 Supplier Thermo Scientific Acros Supplier No. 130971000
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 945-51-7 | C12H10OS | 202.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 945-51-7
Molecular Formula C12H10OS
Molecular Weight (g/mol) 202.27
MDL Number MFCD00002085
InChI Key JJHHIJFTHRNPIK-UHFFFAOYSA-N
Synonym diphenyl sulfoxide, phenyl sulfoxide, sulfinyldibenzene, phenylsulfinylbenzene, diphenyl sulphoxide, sulfoxide, diphenyl, diphenylsulfoxide, benzene, 1,1'-sulfinylbis, 1,1'-sulfinyldibenzene, phenylsulfinyl benzene
PubChem CID 13679
IUPAC Name benzenesulfinylbenzene
SMILES C1=CC=C(C=C1)S(=O)C2=CC=CC=C2

Specifications

Melting Point 66.0°C to 71.0°C
Color White
Boiling Point 206.0°C to 208.0°C (13.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula (C6H5)2SO
Quantity 100 g
Beilstein 06, 300
Fieser 01,348; 17,365
Formula Weight 202.27
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Phenyl sulfoxide, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.