missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetaldehyde dimethyl acetal, 98%

Product Code. 11453008
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11453008

Brand: Thermo Scientific Alfa Aesar A18730.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 101-48-4 | C10H14O2 | 166.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-48-4
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00008487
InChI Key WNJSKZBEWNVKGU-UHFFFAOYSA-N
Synonym 2,2-dimethoxyethyl benzene, phenylacetaldehyde dimethyl acetal, viridine, hyscylene p, 1,1-dimethoxy-2-phenylethane, benzene, 2,2-dimethoxyethyl, phenacetaldehyde dimethyl acetal, padma, 2,2-dimethoxy-1-phenylethane, phenylacetic aldehyde dimethyl acetal
PubChem CID 60995
IUPAC Name 2,2-dimethoxyethylbenzene
SMILES COC(CC1=CC=CC=C1)OC

Specifications

Density 1.004
Boiling Point 220°C
Flash Point 83°C (181°F)
Refractive Index 1.494
Quantity 100 g
Beilstein 879360
Formula Weight 166.22
Percent Purity 98%
Chemical Name or Material Phenylacetaldehyde dimethyl acetal

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.