missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetaldehyde dimethyl acetal, 98%, Thermo Scientific™

Product Code. 10011460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10011460 5 g Glass bottle 5g
10306970 100 g Glass bottle 100g
10705601 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10011460 Supplier Thermo Scientific Acros Supplier No. 130270050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 101-48-4
Molecular Weight (g/mol) 166.22
MDL Number MFCD00008487
InChI Key WNJSKZBEWNVKGU-UHFFFAOYSA-N
Synonym 2,2-dimethoxyethyl benzene, phenylacetaldehyde dimethyl acetal, viridine, hyscylene p, 1,1-dimethoxy-2-phenylethane, benzene, 2,2-dimethoxyethyl, phenacetaldehyde dimethyl acetal, padma, 2,2-dimethoxy-1-phenylethane, phenylacetic aldehyde dimethyl acetal
PubChem CID 60995
IUPAC Name 2,2-dimethoxyethylbenzene
SMILES COC(CC1=CC=CC=C1)OC

Specifications

Acidity 1mg KOH/g max.
Color Colorless to Yellow
Density 1.0040g/mL
Boiling Point 219°C to 221°C (754.0 mmHg)
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C10H14O2
Linear Formula C6H5CH2CH(OCH3)2
Refractive Index 1.4930 to 1.495
Quantity 5 g
Beilstein 07, 293
Specific Gravity 1.004
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in 2 parts of 70% alcohol
Formula Weight 166.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Phenylacetaldehyde dimethyl acetal, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.