missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetyl chloride, 98%

Product Code. 10143660 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10143660 100 g 100g
10021470 5 g 5g
10479630 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10143660 Supplier Thermo Scientific Acros Supplier No. 130301000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 103-80-0 | C8H7ClO | 154.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-80-0
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.59
MDL Number MFCD00000729
InChI Key VMZCDNSFRSVYKQ-UHFFFAOYSA-N
Synonym phenylacetyl chloride, benzeneacetyl chloride, acetyl chloride, phenyl, phenylacetic acid chloride, phenacetyl chloride, phenylacetylchloride, unii-t30899drnd, phenyl acetyl chloride, a-toluoyl chloride, phenylacetyl
PubChem CID 7679
IUPAC Name 2-phenylacetyl chloride
SMILES ClC(=O)CC1=CC=CC=C1

Specifications

Density 1.1600g/mL
Boiling Point 94.0°C to 95.0°C (12.0 mmHg)
Flash Point 102°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2COCl
Refractive Index 1.5320 to 1.5330
Quantity 100 g
Beilstein 09,436
Specific Gravity 1.16
Solubility Information Solubility in water: reacts with water
Formula Weight 154.6
Percent Purity 98%
Chemical Name or Material Phenylacetyl chloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Phenylacetyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Phenylacetyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.