missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetyl Chloride(Benzeneacetyl Chloride), TRC

Product Code. 30343775
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30343775 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30343775 Supplier Toronto Research Chemicals Supplier No. TRCB1888005G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenylacetyl Chloride(Benzeneacetyl Chloride)
CAS 103-80-0
Molecular Formula C8H7ClO
Quantity 5 g
Synonym 2-Phenylacetyl Chloride, 2-Phenylethanoyl Chloride, Phenylacetic Acid Chloride, Phenylacetic Chloride, Phenylacetyl Chloride, alpha-Phenylacetyl Chloride
InChI Formula InChI=1S/C8H7ClO/c9-8(10)6-7-4-2-1-3-5-7/h1-5H,6H2
SMILES ClC(=O)Cc1ccccc1
IUPAC Name 2-phenylacetyl chloride
Molecular Weight (g/mol) 154.59
Formula Weight 154.0185
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.