missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetylene, 98%, pure

Product Code. 10468120 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10468120 25 g Glass bottle 25g
10041830 100 g Glass bottle 100g
11934461 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10468120 Supplier Thermo Scientific Acros Supplier No. 152460250
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 536-74-3 | C8H6 | 102.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536-74-3
Molecular Formula C8H6
Molecular Weight (g/mol) 102.14
InChI Key UEXCJVNBTNXOEH-UHFFFAOYSA-N
Synonym phenylacetylene, phenylethyne, benzene, ethynyl, 1-phenylethyne, phenylacetylide, phenyl acetylene, ethyne, phenyl, acetylene, phenyl, ethynyl-benzene, phenylacethylene
PubChem CID 10821
IUPAC Name ethynylbenzene
SMILES C#CC1=CC=CC=C1

Specifications

Melting Point -44.8°C
Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 142.0°C to 144.0°C
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C≡CH
Refractive Index 1.5470 to 1.5500
Quantity 25 g
Beilstein 05,511
Merck Index 15,391
Specific Gravity 0.93
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,ether and other organic,solvents
Formula Weight 102.14
Percent Purity 98%
Grade Pure
Physical Form Liquid
Chemical Name or Material Phenylacetylene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenylacetylene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.