missing translation for 'onlineSavingsMsg'
Learn More

Phenylbutazone, 98%

Product Code. 11312166
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312166 100 g 100g
11302166 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312166 Supplier Thermo Scientific Alfa Aesar Supplier No. L03449.22
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50-33-9 | C19H20N2O2 | 308.38 g/mol

A non-steroidal anti-inflammatory drug

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-33-9
Molecular Formula C19H20N2O2
Molecular Weight (g/mol) 308.38
MDL Number MFCD00005500
InChI Key VYMDGNCVAMGZFE-UHFFFAOYSA-N
Synonym phenylbutazone, fenilbutazona, butazolidin, diphenylbutazone, phenylbutazonum, butapirazol, butapyrazole, butazolidine, butacote, butadion
PubChem CID 4781
ChEBI CHEBI:48574
IUPAC Name 4-butyl-1,2-diphenylpyrazolidine-3,5-dione
SMILES CCCCC1C(=O)N(N(C1=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 105°C to 108°C
Odor Odorless
Quantity 100 g
UN Number UN2811
Beilstein 290080
Merck Index 14,7277
Formula Weight 308.38
Percent Purity 98%
Chemical Name or Material Phenylbutazone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.