missing translation for 'onlineSavingsMsg'
Learn More

Phenylhydrazine Hydrochloride (1:1), TRC

Product Code. 30416457
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416457 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30416457 Supplier Toronto Research Chemicals Supplier No. TRCP3351101G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenylhydrazine Hydrochloride (1:1)
CAS 59-88-1
Molecular Formula C6 H8 N2 . Cl H
Quantity 1 g
Synonym Hydrazine','phenyl-','hydrochloride (1:1), Hydrazine','phenyl-','monohydrochloride (8CI,9CI), 2-Phenylhydrazinium chloride, N-Phenylhydrazine hydrochloride, Phenylhydrazine hydrochloride, Phenylhydrazine monohydrochloride, Phenylhydrazinium chloride
InChI Formula InChI=1S/C6H8N2.ClH/c7-8-6-4-2-1-3-5-6;/h1-5,8H,7H2;1H
SMILES Cl.NNc1ccccc1
IUPAC Name phenylhydrazine;hydrochloride
Molecular Weight (g/mol) 144.6
Formula Weight 144.0454
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.