missing translation for 'onlineSavingsMsg'
Learn More

Phenylhydrazine Hydrochloride, Fisher Chemical™

Product Code. 12990484 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12990484 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12990484 Supplier Fisher Chemical Supplier No. RP 152 250
  • ONLINE EXCLUSIVE PRICE
    155.00 €
    153.08 € / 250mL
    Save 1.92 €
    1% Off
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

CAS 59-88-1
Molecular Formula C6H9ClN2
MDL Number MFCD00012924
Quantity 250 mL
Synonym phenylhydrazine hydrochloride, phenylhydrazinium chloride, phenylhydrazine-hcl, phenylhydrazine monohydrochloride, phenylhydrazine hcl, hydrazine, phenyl-, monohydrochloride, unii-h7qfk49svd, phenylhydrazin hydrochlorid, phenylhydrazine.hcl, ccris 5707
InChI Key JOVOSQBPPZZESK-UHFFFAOYSA-N
SMILES [H+].[Cl-].NNC1=CC=CC=C1
IUPAC Name hydrogen phenylhydrazine chloride
Molecular Weight (g/mol) 144.60
PubChem CID 60962
compliance-icons
Product Identifier
  • Phenylhydrazine hydrochloride solution. European Pharmacopoeia Ref: 1064501 (RP 152)
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.