missing translation for 'onlineSavingsMsg'
Learn More

Phenylmagnesium bromide, 3M in ether

Product Code. 15482967
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15482967 500 mL 500mL
15472967 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15482967 Supplier Thermo Scientific Alfa Aesar Supplier No. 087326.AP
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Grignard reagent | CAS: 100-58-3 | C6H5BrMg | 181.32 g/mol

Grignard reagent Phenylmagnesium bromide is an important reagent called as Grignard reagent used in the synthesis of organic and oraganometallic reactions. It is used to prepare phenyl aryl sulphides, sulphoxides, and sulphones. It acts as a strong acid and Lewis acid and undergoes addition reaction with many unsaturated functional groups. It is involved in the synthesis of end-functionalized regioregular poly(3-alkylthiophene)s, 1,3,3-trimethyl-6-phenyl-2-oxabicyclo[2.2.2]octan-6-ol, o-substituted benzophenones and 6-benzyl-1,3,3-trimethyl-2-oxabicyclo[2.2.2]octan-6-ol. In addition to this, it is used for the monalkylation of primary amines to get secondary amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-58-3
Molecular Formula C6H5BrMg
Molecular Weight (g/mol) 181.32
MDL Number MFCD00000038
InChI Key ANRQGKOBLBYXFM-UHFFFAOYSA-M
Synonym phenylmagnesium bromide, bromophenylmagnesium, magnesium, bromophenyl, phmgbr, phenylmagnesiumbromide, bromo phenyl magnesium, phenyl magnesium bromide, phenylmagnesium bromide solution, 3.0 m in diethyl ether, bromomagnesiobenzene, phenyhnagnesium bromide, grignard reagent
PubChem CID 66852
SMILES Br[Mg]C1=CC=CC=C1

Specifications

Color Yellow to Brown
Density 1.14 g/mL
Odor Ether-like
Quantity 500 mL
UN Number UN3399
Sensitivity Air, light, and moisture sensitive; May form a Precipitate
Solubility Information Miscible with terahydrofuran.
IUPAC Name magnesium;benzene;bromide
Formula Weight 181.31
Concentration or Composition (by Analyte or Components) 3M in ether
Physical Form Liquid
Chemical Name or Material Phenylmagnesium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.