missing translation for 'onlineSavingsMsg'
Learn More

Phenylmaleic anhydride, 97%

Product Code. 11372166
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372166 5 g 5g
11362166 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372166 Supplier Thermo Scientific Alfa Aesar Supplier No. L03453.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 36122-35-7 | C10H6O3 | 174.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36122-35-7
Molecular Formula C10H6O3
Molecular Weight (g/mol) 174.16
MDL Number MFCD00015472
InChI Key QZYCWJVSPFQUQC-UHFFFAOYSA-N
Synonym phenylmaleic anhydride, 3-phenyl-furan-2,5-dione, 3-phenyl-2,5-furandione, 3-phenyl-2,5-dihydrofuran-2,5-dione, 2,5-furandione, 3-phenyl, phenylmaleicanhydride, phenylmalecanhydrde, acmc-20ajwi, phenyl maleic anhydride, phenyl-maleic anhydride
PubChem CID 99174
IUPAC Name 3-phenylfuran-2,5-dione
SMILES O=C1OC(=O)C(=C1)C1=CC=CC=C1

Specifications

Melting Point 118°C to 119°C
Quantity 5 g
Beilstein 127301
Sensitivity Moisture sensitive
Formula Weight 174.16
Percent Purity 97%
Chemical Name or Material Phenylmaleic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.