missing translation for 'onlineSavingsMsg'
Learn More

Phenylmercuric Chloride, TRC

Product Code. 30418412
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30418412 5 g 5g
30421241 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30418412 Supplier Toronto Research Chemicals Supplier No. TRCP3356585G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenyl-mercury-chloride
CAS 100-56-1
Molecular Formula C6 H5 Cl Hg
Quantity 5 g
Synonym Phenylmercury Chloride, Mercury','chlorophenyl-, Phenylmercury chloride (6CI), Chlorophenylmercury, Agrenal, Agronal, Benzene','(chloromercurio)-, Hexason, Mercutal, Merfazin, Mersolite 2, Monophenylmercury chloride, PMC, PMC (fungicide), Phenyl chloromercury, Phenylmercuric chloride, Phenylmercury(II) chloride
InChI Formula InChI=1S/C6H5.ClH.Hg/c1-2-4-6-5-3-1;;/h1-5H;1H;/q;;+1/p-1
SMILES Cl[Hg]c1ccccc1
IUPAC Name chloro(phenyl)mercury
Molecular Weight (g/mol) 313.15
Formula Weight 313.9786
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.