missing translation for 'onlineSavingsMsg'
Learn More

Phenylmercuric Chloride, TRC

Product Code. 30421241
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30421241 1 g 1g
30418412 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30421241 Supplier Toronto Research Chemicals Supplier No. TRCP3356581G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenyl-mercury-chloride
CAS 100-56-1
Molecular Formula C6 H5 Cl Hg
Quantity 1 g
Synonym Phenylmercury Chloride, Mercury','chlorophenyl-, Phenylmercury chloride (6CI), Chlorophenylmercury, Agrenal, Agronal, Benzene','(chloromercurio)-, Hexason, Mercutal, Merfazin, Mersolite 2, Monophenylmercury chloride, PMC, PMC (fungicide), Phenyl chloromercury, Phenylmercuric chloride, Phenylmercury(II) chloride
InChI Formula InChI=1S/C6H5.ClH.Hg/c1-2-4-6-5-3-1;;/h1-5H;1H;/q;;+1/p-1
SMILES Cl[Hg]c1ccccc1
IUPAC Name chloro(phenyl)mercury
Molecular Weight (g/mol) 313.15
Formula Weight 313.9786
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.