missing translation for 'onlineSavingsMsg'
Learn More

Phenylmercury(II) chloride, 96%, Hg 63.5%

Product Code. 11314287
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314287 10 g 10g
11324287 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11314287 Supplier Thermo Scientific Alfa Aesar Supplier No. 037113.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-56-1 | C6H5ClHg | 313.15 g/mol

The reaction of phenylmercury (II) chloride with acetophenonethiosemicarbazone (Hatsc) in ethanol in 1: 1 mole ratio undergoes symmetrisation forming the products, HgCl2 (Hatse) 2 and Ph2Hg instead of the anticipated compound PhHgCl (Hatse). A new method for the determination of chloride ion is based on the formation of phenylmercury(II) chloride, its extraction into chloroform and reaction with sodium diethyldithiocarbamate to form phenylmercury(II) diethyldithiocarbamate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-56-1
Molecular Formula C6H5ClHg
Molecular Weight (g/mol) 313.15
MDL Number MFCD00000463
InChI Key AWGTVRDHKJQFAX-UHFFFAOYSA-M
Synonym phenylmercuric chloride, agrenal, phenylmercury chloride, mercury, chlorophenyl, hexason, mercutal, merfazin, stopspot, agronal, mersolite 2
PubChem CID 7511
IUPAC Name chloro(phenyl)mercury
SMILES Cl[Hg]C1=CC=CC=C1

Specifications

Melting Point 246°C to 250°C
Color White
Quantity 10 g
UN Number UN2026
Merck Index 14,7301
Solubility Information Soluble in benzene, ether, pyridine; Very slightly soluble in water
Formula Weight 313.18
Percent Purity 96%
Concentration or Composition (by Analyte or Components) Hg 63.5%
Physical Form Powder
Chemical Name or Material Phenylmercury(II) chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.