missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenylmethanesulfonyl fluoride, 99%

Product Code. 10358110 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10358110 1 g Glass bottle 1g
10042680 5 g Glass bottle 5g
10105180 10 g Glass bottle 10g
10267023 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10358110 Supplier Thermo Scientific Chemicals Supplier No. 215740010
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 329-98-6 | C7H7FO2S | 174.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 329-98-6
Molecular Formula C7H7FO2S
Molecular Weight (g/mol) 174.19
MDL Number MFCD00007424
InChI Key YBYRMVIVWMBXKQ-UHFFFAOYSA-N
Synonym phenylmethylsulfonyl fluoride, pmsf, benzenemethanesulfonyl fluoride, benzylsulfonyl fluoride, alpha-toluenesulfonyl fluoride, benzylsulphonyl fluoride, alpha-toluenesulphonyl fluoride, phenylmethylsulfonylfluoride, phenylmethylsulphonyl fluoride, .alpha.-toluenesulfonyl fluoride
PubChem CID 4784
ChEBI CHEBI:8102
IUPAC Name phenylmethanesulfonyl fluoride
SMILES C1=CC=C(C=C1)CS(=O)(=O)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 90.0°C to 94.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2SO2F
Quantity 1 g
Beilstein 11, III, 331
Merck Index 15, 7659
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in xylene and petroleum ether
Formula Weight 174.19
Percent Purity 99%
Physical Form Needles (May Agglomerate)
Chemical Name or Material Phenylmethanesulfonyl fluoride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phenylmethanesulfonyl fluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.