missing translation for 'onlineSavingsMsg'
Learn More

Phenylmethylsulfonyl Fluoride, TRC

Product Code. 30425223
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30425223 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30425223 Supplier Toronto Research Chemicals Supplier No. TRCP33754510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenylmethylsulfonyl Fluoride
CAS 329-98-6
Molecular Formula C7H7FO2S
Quantity 10 g
Synonym α-Toluenesulfonyl Fluoride, Benzylsulfonyl Fluoride, NSC 88499, PMSF, Phenylmethanesulfonyl Fluoride
InChI Formula InChI=1S/C7H7FO2S/c8-11(9,10)6-7-4-2-1-3-5-7/h1-5H,6H2
SMILES C1=CC=C(C=C1)CS(=O)(=O)F
IUPAC Name phenylmethanesulfonyl fluoride
Molecular Weight (g/mol) 174.19
Formula Weight 174.0151
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.