missing translation for 'onlineSavingsMsg'
Learn More

Phenylselenenyl chloride, 98%

Product Code. 11441910
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441910 25 g 25g
11431910 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441910 Supplier Thermo Scientific Alfa Aesar Supplier No. A12751.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5707-04-0 | C6H5ClSe | 191.53 g/mol

Phenylselenenyl chloride is used in the preparation of (cyclobut-1-enylselanyl)benzene. It is an important versatile reagent used in organic synthesis. Further, it serves as a useful synthon for 2-amino alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylselenenyl chloride is used in the preparation of (cyclobut-1-enylselanyl)benzene. It is an important versatile reagent used in organic synthesis. Further, it serves as a useful synthon for 2-amino alcohols.

Solubility
Soluble in methanol.

Notes
Moisture sensitive. Incompatible with strong bases, strong oxidizing agents and strong alkalis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5707-04-0
Molecular Formula C6H5ClSe
Molecular Weight (g/mol) 191.53
MDL Number MFCD00000478
InChI Key WJCXADMLESSGRI-UHFFFAOYSA-N
Synonym phenylselenenyl chloride, phenylselenyl chloride, benzeneselenenyl chloride, chloroselenobenzene, phenyl hypochloroselenoite, phenylselenium chloride, phsecl, phenylselenenylchloride, phenylselanyl chlorane, chloroselanylbenzene
PubChem CID 21928
IUPAC Name phenyl selenohypochlorite
SMILES Cl[Se]C1=CC=CC=C1

Specifications

Melting Point 60°C to 65°C
Boiling Point 120°C (20 mmHg)
Odor Stench
Quantity 25 g
UN Number UN2928
Beilstein 1237091
Sensitivity Air and moisture sensitive
Solubility Information Soluble in methanol.
Formula Weight 191.52
Percent Purity 98%
Chemical Name or Material Phenylselenenyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.