missing translation for 'onlineSavingsMsg'
Learn More

Phenylseleninic Anhydride, TRC

Product Code. 30339782
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30339782 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30339782 Supplier Toronto Research Chemicals Supplier No. TRCB398190100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenylseleninic Anhydride
CAS 17697-12-0
Molecular Formula C12 H10 O3 Se2
Quantity 100 mg
Synonym Phenylseleninic Anhydride
InChI Formula InChI=1S/C12H10O3Se2/c13-16(11-7-3-1-4-8-11)15-17(14)12-9-5-2-6-10-12/h1-10H
SMILES O=[Se](O[Se](=O)c1ccccc1)c2ccccc2
IUPAC Name phenylseleninyl benzeneseleninate
Molecular Weight (g/mol) 360.126
Formula Weight 361.896
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.