missing translation for 'onlineSavingsMsg'
Learn More

Phenylselenyl bromide, 98%

Product Code. 10328120 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10328120 5 g Glass bottle 5g
10713421 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10328120 Supplier Thermo Scientific Acros Supplier No. 223060050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 34837-55-3 | C6H5BrSe | 235.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34837-55-3
Molecular Formula C6H5BrSe
Molecular Weight (g/mol) 235.97
MDL Number MFCD00000047
InChI Key LCEFEIBEOBPPSJ-UHFFFAOYSA-N
Synonym phenylselenyl bromide, benzeneselenenyl bromide, phenylselenenyl bromide, bromoselenobenzene, phsebr, phenylselenylbromide, benzeneselenyl bromide, phenylselanyl bromane, brseph, phenylseleno bromide
PubChem CID 123446
IUPAC Name phenyl selenohypobromite
SMILES C1=CC=C(C=C1)[Se]Br

Specifications

Melting Point 60°C to 62°C
Color Black to Brown
Boiling Point 107°C to 108°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5SeBr
Quantity 5 g
Beilstein 06, II, 319
Fieser 06,459; 07,286; 11,33; 13,26; 01,1047; 11,487; 16,272
Solubility Information Solubility in water: insoluble
Formula Weight 235.97
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material Phenylselenyl bromide, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.