missing translation for 'onlineSavingsMsg'
Learn More

Phenylselenyl chloride, 98%

Product Code. 10083443 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10083443 1 g Glass bottle 1g
10347740 10 g Glass bottle 10g
10672023 25 g Glass bottle 25g
10201293 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10083443 Supplier Thermo Scientific Acros Supplier No. 179510010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5707-04-0 | C6H5ClSe | 191.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5707-04-0
Molecular Formula C6H5ClSe
Molecular Weight (g/mol) 191.53
MDL Number MFCD00000478
InChI Key WJCXADMLESSGRI-UHFFFAOYSA-N
Synonym phenylselenenyl chloride, phenylselenyl chloride, benzeneselenenyl chloride, chloroselenobenzene, phenyl hypochloroselenoite, phenylselenium chloride, phsecl, phenylselenenylchloride, phenylselanyl chlorane, chloroselanylbenzene
PubChem CID 21928
IUPAC Name phenyl selenohypochlorite
SMILES Cl[Se]C1=CC=CC=C1

Specifications

Melting Point 61.0°C to 65.0°C
Color Orange
Boiling Point 120.0°C (20.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula C6H5SeCl
Quantity 1 g
Beilstein 06, III, 1110
Fieser 05,518; 06,459; 07,286; 09,25; 11,33; 13,26; 16,19; 17,26
Formula Weight 191.52
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Phenylselenyl chloride, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenylselenyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.