missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrichlorosilane, 95%

Product Code. 10438100 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
10mL
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10438100 10 mL Glass bottle 10mL
10635952 250 mL Glass bottle 250mL
10035723 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10438100 Supplier Thermo Scientific Acros Supplier No. 131000100
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 98-13-5 | C6H5Cl3Si | 211.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-13-5
Molecular Formula C6H5Cl3Si
Molecular Weight (g/mol) 211.55
MDL Number MFCD00000479
InChI Key ORVMIVQULIKXCP-UHFFFAOYSA-N
Synonym trichloro phenyl silane, phenyltrichlorosilane, silane, trichlorophenyl, phenyl trichlorosilane, phenylsilicon trichloride, silane, phenyltrichloro, silicon phenyl trichloride, benzene, trichlorosilyl, unii-m7199jl06m, phenyl-trichloro-silane
PubChem CID 7372
IUPAC Name trichloro(phenyl)silane
SMILES C1=CC=C(C=C1)[Si](Cl)(Cl)Cl

Specifications

Color Colorless to Yellow
Density 1.3210g/mL
Boiling Point 201°C
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C6H5SiCl3
Refractive Index 1.5220 to 1.524
Quantity 10 mL
Beilstein 16, 911
Specific Gravity 1.321
Solubility Information Solubility in water: reacts. Other solubilities: soluble in benzene,ehter and perchloroethylene
Formula Weight 211.55
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Phenyltrichlorosilane, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phenyltrichlorosilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • EUH014-Reacts violently with water.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P232-Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.