missing translation for 'onlineSavingsMsg'
Learn More

Phenyltriethoxysilane, 98%

Product Code. 10751451 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10751451 1 kg Glass bottle 1kg
10634672 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10751451 Supplier Thermo Scientific Acros Supplier No. 174640010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 780-69-8 | C12H20O3Si | 240.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 780-69-8
Molecular Formula C12H20O3Si
Molecular Weight (g/mol) 240.37
MDL Number MFCD00009065
InChI Key JCVQKRGIASEUKR-UHFFFAOYSA-N
Synonym phenyltriethoxysilane, triethoxy phenyl silane, silane, triethoxyphenyl, triethoxyfenylsilan, silane, phenyltriethoxy, benzeneorthosiliconic acid, triethyl ester, phenyl triethoxysilane, benzene, triethoxysilyl, triethoxyfenylsilan czech, unii-qi310t2x15
PubChem CID 13075
IUPAC Name triethoxy(phenyl)silane
SMILES CCO[Si](OCC)(OCC)C1=CC=CC=C1

Specifications

Color Colorless
Density 0.9960g/mL
Boiling Point 235.0°C
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5Si(OC2H5)3
Refractive Index 1.4594 to 1.4614
Quantity 1 kg
Beilstein 16, 911
Specific Gravity 0.996
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 240.38
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Phenyltriethoxysilane, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phenyltriethoxysilane
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.