missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrimethylammonium chloride, 98+%

Product Code. 11449887
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449887 100 g 100g
11459887 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449887 Supplier Thermo Scientific Alfa Aesar Supplier No. B22851.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 138-24-9 | C9H14ClN | 171.67 g/mol

Phenyltrimethylammonium chloride is an effective phase transfer catalyst, methylating agent, also useful for alkaline depolymerizations, corrosion inhibitor for low carbon steel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyltrimethylammonium chloride is an effective phase transfer catalyst, methylating agent, also useful for alkaline depolymerizations, corrosion inhibitor for low carbon steel.

Solubility
Soluble in water 333 g/L.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agent, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138-24-9
Molecular Formula C9H14ClN
Molecular Weight (g/mol) 171.67
MDL Number MFCD00011790
InChI Key MQAYPFVXSPHGJM-UHFFFAOYSA-M
Synonym phenyltrimethylammonium chloride, n,n,n-trimethylbenzenaminium chloride, trimethylphenylammonium chloride, n,n,n-trimethylanilinium chloride, ammonyx 200, benzenaminium, n,n,n-trimethyl-, chloride, trimethylphenylammoniumchloride, trimethylanilinium chloride, ammonium, phenyltrimethyl-, chloride, trimethylphenyl ammonium chloride
PubChem CID 67309
SMILES [Cl-].C[N+](C)(C)C1=CC=CC=C1

Specifications

Melting Point ∼236°C (sublimation)
Quantity 100 g
UN Number UN2811
Beilstein 3572527
Sensitivity Hygroscopic
Solubility Information Soluble in water 333g/L.
IUPAC Name N,N,N-trimethylanilinium chloride
Formula Weight 171.67
Percent Purity ≥98%
Chemical Name or Material Phenyltrimethylammonium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.