missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrimethylammonium iodide, 99%, Thermo Scientific™

Product Code. 10646942 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
100g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10646942 100g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10646942 Supplier Thermo Scientific Acros Supplier No. 160681000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifications

CAS 98-04-4
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Molecular Formula C9H14IN
Linear Formula C6H5N(CH3)3I
MDL Number MFCD00011791
Quantity 100g
Merck Index 15, 7428
Synonym phenyltrimethylammonium iodide, trimethylphenylammonium iodide, n,n,n-trimethylanilinium iodide, trimethylanilinium iodide, trimethylanillinium iodide, n,n-dimethylaniline methiodide, unii-4o888xq7fp, ammonium, phenyltrimethyl-, iodide, n,n,n-trimethylbenzenaminium iodide, benzenaminium, n,n,n-trimethyl-, iodide
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, insoluble in chloroform
InChI Key KKLAORVGAKUOPZ-UHFFFAOYSA-M
SMILES C[N+](C)(C)C1=CC=CC=C1.[I-]
IUPAC Name trimethyl(phenyl)azanium;iodide
Molecular Weight (g/mol) 263.11
PubChem CID 94135
Formula Weight 263.11
Percent Purity 99%
Chemical Name or Material Phenyltrimethylammonium iodide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Phenyltrimethylammonium iodide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.