missing translation for 'onlineSavingsMsg'
Learn More

Phenylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Product Code. 15425395
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15425395 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15425395 Supplier Thermo Scientific Alfa Aesar Supplier No. H58659.AD
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 38111-44-3 | C6H5BrZn | 222.39 g/mol

It is used in the cross-coupling reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the cross-coupling reaction.

Solubility
Reacts with water.

Notes
Handle under dry protective gas. Air sensitive. Store away from strong bases and oxidizing agents, heat, acids and wate
TRUSTED_SUSTAINABILITY

Specifications

CAS 38111-44-3
Density 0.99
Molecular Formula C6H5BrZn
MDL Number MFCD01319861
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Synonym phenylzinc bromide, zinc, bromophenyl, phenylzinc bromide solution, benzene,bromozinc 1+, phenylzinc bromide 0.5 m in tetrahydrofuran, phenylzinc bromide solution, 0.5 m in thf, phenylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
Solubility Information Reacts with water.
InChI Key HMFWJMIDCMEJHO-UHFFFAOYSA-M
SMILES [Zn++].[Br-].C1=CC=[C-]C=C1
IUPAC Name zinc;benzene;bromide
Molecular Weight (g/mol) 222.39
PubChem CID 16213817
Formula Weight 222.39
Chemical Name or Material Phenylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.