missing translation for 'onlineSavingsMsg'
Learn More

uvex Pheos B-S-WR Safety Helmet

Product Code. 17514116 Shop All Uvex Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17514116 White Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17514116 Supplier uvex Supplier No. 9772031
  • ONLINE EXCLUSIVE PRICE
    457.00 €
    428.65 € / Pack of 25
    Save 28.35 €
    6% Off
View all promotions
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lightweight safety helmets that feature a sporty design and offer maximum wearer comfort.

  • Lightweight helmet in a sporty design
  • Three variable air vents, one ventilation zone on the forehead and two at the back, for perfect air circulation
  • Adapter systems for uvex visors, uvex earmuffs, and helmet lamps
  • Shorter brim for a wider field of upward vision
  • 6-point textile suspension harness with rotating size adjusters for individual width adjustment
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 397
Color White
For Use With (Equipment) Hard Hats
Material HDPE
Model 9772
Product Type Industrial Safety Helmet
Size 52 to 61 cm
Size (Metric) 52 to 61 cm
Style Short Brim
Ventilated Ventilated
Head Style Front Brim
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.