Learn More
uvex Pheos Eyewear
Product Code. 17767689 Shop All Uvex Products
Change view
Click to view available options
Frame Color: Black and gray ; Coating: supravision excellence
Product Code.
17767689 Frame Color: Black and gray ; Coating: supravision excellence
1 options
Product Code. 17767689 Supplier uvex Supplier No. 9192485
  • ONLINE EXCLUSIVE PRICE
    12.40 €
    11.50 € / Pack of 1
    Save 0.90 €
    7% Off
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
More than just a common spectacle - the modern fashion-look makes it a welcome companion at everyday work
  • Perfect all-round protection, large field of vision through duo spherical lens, outstanding ventilation thanks to integrated uvex climazone temple concept
  • Coating uvex supravision excellence
  • TRUSTED_SUSTAINABILITY
    Specifications
    Lens Type Chemical-resistant
    Lens Coating Anti-fog, Scratch-resistant
    Color Gray, Black
    Certifications/Compliance (Europe) EN 166, EN 170
    Lens Tint Clear
    Product Type Ergonomic Safety Glasses
    Material Polycarbonate
    Disposable Reusable
    Lens Material Polycarbonate
    Frame Material Plastic
    UV Protection UV400
    Frame Type Standard