missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phire Hot Start II DNA Polymerase

Product Code. 10247467 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Colorless
Green
No. of Reactions:
1000 Reactions
200 Reactions
Unit Size:
1000 reactions
200 reactions
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color No. of Reactions unitSize
10247467 Colorless 1000 Reactions 1000 reactions
10185667 Colorless 200 Reactions 200 reactions
13565160 Green 1000 Reactions 1000 reactions
13555160 Green 200 Reactions 200 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price
Product Code. 10247467 Supplier Thermo Scientific™ Supplier No. F122L

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price

Thermo Scientific Phire Hot Start II DNA Polymerase is an enhanced PCR enzyme for routine and high throughput PCR applications. It is significantly faster, extremely robust, and also capable of amplifying long DNA fragments with high yields.

Thermo Scientific Phire Hot Start II DNA Polymerase is an enhanced PCR enzyme for routine and high throughput PCR applications. It outperforms every Taq-based hot start polymerase on the market. Phire Hot Start II DNA Polymerase is significantly faster, extremely robust, and also capable of amplifying long DNA fragments with high yields. These features are achieved through advanced protein engineering of the polymerase. It incorporates a unique double-stranded DNA binding domain which allows short extension times (10–15 s/kb), improves yields, and increases fidelity 2-fold compared to Taq DNA polymerase (Cat. No. EP0072).

The hot start modification of Phire Hot Start II DNA Polymerase is based on the Affibody inactivation technology. This technology increases the specificity of the PCR reaction with no additional time required for initial activation of the enzyme.

Phire Green format is a combination of Phire Hot Start II DNA Polymerase and 5X Phire Green Buffer. The buffer includes a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with enzyme performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.

Features

  • No reactivation step with hot start
  • Amplification four times faster than hot start Taq
  • Minimal reaction optimization due to high inhibitor tolerance
  • Abundant products due to high efficiency
  • Amplifies significantly longer DNA fragments than hot start Taq
  • Direct loading on gel with Green Buffer

Applications

  • Hot Start PCR
  • Standard PCR
  • Non-high-fidelity PCR
  • Fast PCR
  • High throughput PCR
  • Genotyping

Notes

  • The optimal annealing temperature for Phire DNA Polymerases may differ significantly from that of Taq-based polymerases.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Phire Hot Start II DNA Polymerase, 1 x 1 mL
• 5X Phire Reaction Buffer, 7 x 1.5 mL
• DMSO, 1 x 500 μL

Store at -20°C.
GC-Rich PCR Performance Medium
Polymerase Phire Hot Start II DNA Polymerase
Reaction Speed Fast
Product Type DNA Polymerase
Purity or Quality Grade DHPLC
Quantity 1,000 reactions
Shipping Condition Wet or Dry Ice
For Use With (Application) Cloning, Hot-start PCR
Fidelity (vs. Taq) 2X
Hot Start Built-In Hot Start
No. of Reactions 1000 Reactions
Overhang Blunt
Reaction Format Separate Components
Starting Material DNA
Color Colorless
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.