Learn More
Thermo Scientific™ Phire Plant Direct PCR Master Mix
Amplify DNA directly from plant samples without prior DNA purification with Phire Plant Direct PCR Master Mix
Brand: Thermo Scientific™ F160L
601.73 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.18000kg
Description
Thermo Scientific™ Phire Plant Direct PCR Master Mix is designed to amplify DNA directly from plant samples. No DNA purification is required prior to PCR. The kit is based on Phire Hot Start II DNA Polymerase, a specially engineered enzyme with a DNA binding domain. The unique features of this DNA polymerase make it extremely robust and tolerant of many PCR inhibitors present in plant material like complex polysaccharides, polyphenols and others. Phire Plant Direct PCR Master Mix has been tested with leaves and seeds from a wide variety of plant species. The kit includes a complete set of optimized reagents, detailed protocols making it an ideal choice for amplification of plant DNA. When combined with Thermo Scientific™ Arktik™ Thermal Cycler, PCR protocols can be completed in as little as 40 minutes.
Highlights- Direct PCR - sample is added directly to PCR reaction, therefore there is no need for time-consuming and expensive DNA purification steps.
- Dilution & Storage protocol available - allows tens of PCR reactions from one tiny sample and allows re-testing
- Specially engineered Phire Hot Start II DNA polymerase - extremely short PCR protocol times
- Master Mix format with premixed gel loading dye - minimizes possibility of cross-contamination, reduces sample handling and allows direct loading on gel
- Genotyping
- Transgene detection
- Knockout analysis
- Sequencing
- 2x Phire Plant Direct PCR Master Mix
- Dilution Buffer
- Control Primer Mix (25 μM each)
- Water, nuclease- free
- O'GeneRuler Express DNA Ladder
Specifications
Contains: • 10 x 1.25 mL Phire Plant Direct PCR Master Mix (2X) • 2 x 5 mL Dilution Buffer • 40 μL Control Primer Mix (25 μM each) • 10 x 1.25 mL Water, nuclease- free • 50 μg O'GeneRuler Express DNA Ladder (sufficient for 100 applications) Store at -20°C. |
|
Direct PCR | |
Fast | |
Direct PCR Master Mix | |
Genotyping | |
1250 x 50 L Reactions | |
SuperMix or Master Mix | |
DNA |
Medium | |
Phire Hot Start II DNA polymerase | |
Phire | |
1250 x 50 μL Reactions | |
Built-In Hot Start | |
Blunt | |
Plant |
For Research Use Only. Not for use in diagnostic procedures.