missing translation for 'onlineSavingsMsg'
Learn More

Phloroglucinol dihydrate, 98%

Product Code. 11459963
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459963 500 g 500g
11439963 25 g 25g
11449963 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11459963 Lieferant Thermo Scientific Alfa Aesar Lieferanten-Nr. A13034.36
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 6099-90-7 | C6H10O5 | 162.141 g/mol

Phloroglucinol dihydrate is utilized in the preparation of active pharmaceutical ingredients such as flopropione. It acts as a reagent for the Tollens’ test for pentoses. It is mixed with hydrochloric acid and plays an important role for the detection of lignin (Wiesner test). It is involved in the analysis and qualitative detection of free hydrochloric acid in gastric juice by using Gunzburg reagent. Further, it serves as a coupling agent in printing. It is also employed as a reagent for pentoses, pentosans, aldehydes and as a decalcifier of bone specimens in microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phloroglucinol dihydrate is utilized in the preparation of active pharmaceutical ingredients such as flopropione. It acts as a reagent for the Tollens’ test for pentoses. It is mixed with hydrochloric acid and plays an important role for the detection of lignin (Wiesner test). It is involved in the analysis and qualitative detection of free hydrochloric acid in gastric juice by using Gunzburg reagent. Further, it serves as a coupling agent in printing. It is also employed as a reagent for pentoses, pentosans, aldehydes and as a decalcifier of bone specimens in microscopy.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with acid chlorides, acid anhydrides, bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 6099-90-7
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.141
MDL Number MFCD00149090
InChI Key MPYXTIHPALVENR-UHFFFAOYSA-N
Synonym phloroglucinol dihydrate, benzene-1,3,5-triol dihydrate, 1,3,5-trihydroxybenzene dihydrate, 1,3,5-benzenetriol, dihydrate, phloroglucinol, dihydrate, sym-trihydroxybenze, unii-0x7b22brg1, ccris 6521, 1,3,5-benzoltrioldihydrat, phloroglucinoldihydrate
PubChem CID 80196
IUPAC Name benzene-1,3,5-triol;dihydrate
SMILES C1=C(C=C(C=C1O)O)O.O.O

Spezifikation

Melting Point 214°C to 217°C
Odor Odorless
Quantity 500 g
Sensitivity Light sensitive
Merck Index 14,7328
Solubility Information Slightly soluble in water.
Formula Weight 162.14 (126.11 Anhydrous)
Percent Purity 98%
Chemical Name or Material Phloroglucinol dihydrate

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.