missing translation for 'onlineSavingsMsg'
Learn More

Phloroglucinol, TRC

Product Code. 30422537
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30422537 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30422537 Supplier Toronto Research Chemicals Supplier No. TRCP34000025G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phloroglucinol
CAS 108-73-6
Molecular Formula C6 H6 O3
Quantity 25 g
Synonym Phloroglucinol anhydrous, Benzene-1,3,5-triol, Phloroglucinol (8CI), 1,3,5-Trihydroxybenzene, 3,5-Dihydroxyphenol, Benzene-s-triol, Dilospan S, NSC 1572, Phloroglucin, Phloroglucine, Spasfon-Lyoc, sym-Trihydroxybenzene
InChI Formula InChI=1S/C6H6O3/c7-4-1-5(8)3-6(9)2-4/h1-3,7-9H
SMILES Oc1cc(O)cc(O)c1
IUPAC Name benzene-1,3,5-triol
Molecular Weight (g/mol) 126.11
Formula Weight 126.0317
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.