missing translation for 'onlineSavingsMsg'
Learn More

Phloxine B, Certified, 85%, Thermo Scientific™

Product Code. 10445661 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10445661 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10445661 Supplier Thermo Scientific Acros Supplier No. 417870250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 18472-87-2
Absorption Ratio 1.38 to 2.24
Dye Content 80% min.
Infrared Spectrum Authentic
Lambda Max 546 to 550nm
Packaging Glass bottle
Assay Percent Range 85%
Molecular Formula C20 H2 Br4 Cl4 Na2 O5
MDL Number MFCD00005061
Beilstein 19, IV, 2921
Merck Index 15, 7440
Synonym Acid Red 92, C.I. 45410
Color Brown/Red to Red
Quantity 25 g
Physical Form Powder
Formula Weight 829.64
Grade Pure
Chemical Name or Material Phloxine B
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.