missing translation for 'onlineSavingsMsg'
Learn More

Phloxine B, TRC

Product Code. 30136557
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136557 10 g 10g
30133531 25 g 25g
30006290 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136557 Supplier Toronto Research Chemicals Supplier No. TRCP34000810G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18472-87-2
Molecular Formula C20 H2 Br4 Cl4 O5 . 2 Na
Molecular Weight (g/mol) 829.63
Synonym Spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one, 2',4',5',7'-tetrabromo-4,5,6,7-tetrachloro-3',6'-dihydroxy-, sodium salt (1:2), Fluorescein, 2',4',5',7'-tetrabromo-4,5,6,7-tetrachloro-, disodium salt (8CI), Phloxin B (6CI), Spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one, 2',4',5',7'-tetrabromo-4,5,6,7-tetrachloro-3',6'-dihydroxy-, disodium salt (9CI), 11969 Red, 3427 Veri Pur Pink, Acid Red 92, Aizen Acid Phloxine PB, C.I. 45410, C.I. Acid Red 92, Cyanosin, Cyanosin (acid dye), Cyanosin B, Cyanosine, D and C Red No. 28, D&C Red 28, D&C Red 28-308267, D&C Red No. 28, D&C Red No. 28-15347, D&C Red No. 28-38015, Daiwa Red 104WB, Daiwa Red 45, DermaGlo DG-R 428, Disodium 9-(3',4',5',6'-tetrachloro-o-carboxyphenyl)-6-hydroxy-6-hydroxy-2,4,5,7-tetrabromo-3-isoxanthone, Eosin blue, Eosin bluish, Eosine blue, Eosine bluish, Food Red 104, Food Red No. 104, Japan Red 104, Japan Red 104-1, Japan Red No. 104-1, Orient Water Pink 2, Phloxine B, Phloxine B 38015, Phloxine P, Phyloxine B, Red 104, Red No. 104, Red No.104-1, Triacid Floxine 2G, Water Pink 2
IUPAC Name disodium;2',4',5',7'-tetrabromo-4,5,6,7-tetrachloro-3-oxospiro[2-benzofuran-1,9'-xanthene]-3',6'-diolate
SMILES [Na+].[Na+].[O-]c1c(Br)cc2c(Oc3c(Br)c([O-])c(Br)cc3C24OC(=O)c5c(Cl)c(Cl)c(Cl)c(Cl)c45)c1Br

Specifications

Quantity 10 g
InChI Formula InChI=1S/C20H4Br4Cl4O5.2Na/c21-5-1-3-17(9(23)15(5)29)32-18-4(2-6(22)16(30)10(18)24)20(3)8-7(19(31)33-20)11(25)13(27)14(28)12(8)26;;/h1-2,29-30H;;/q;2*+1/p-2
Formula Weight 823.5185
Chemical Name or Material Phloxine B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.