missing translation for 'onlineSavingsMsg'
Learn More

Phorate, 85%, TRC

Product Code. 30041156
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041156 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30041156 Supplier Toronto Research Chemicals Supplier No. TRCP353500100MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 298-02-2
Molecular Formula C7 H17 O2 P S3
Quantity 100 mg
Synonym Phosphorodithioic acid, O,O-diethyl S-[(ethylthio)methyl] ester, American Cyanamid 3911, ENT 24,042, Experimental Insecticide 3911, Granutox, L 11/6, O,O-Diethyl S-[(ethylthio)methyl] phosphorodithioate, O,O-Diethyl S-ethylmercaptomethyl dithiophosphate, O,O-Diethyl S-ethylthiomethyl dithiophosphate, Phorate, Phorate 10G, Thimet, Thimet 10G, Thimet G, Timet, VUAgT 182, suScon FuMing
InChI Formula InChI=1S/C7H17O2PS3/c1-4-8-10(11,9-5-2)13-7-12-6-3/h4-7H2,1-3H3
SMILES CCOP(=S)(OCC)SCSCC
IUPAC Name diethoxy-(ethylsulfanylmethylsulfanyl)-sulfanylidene-λ^{5}-phosphane
Molecular Weight (g/mol) 260.38
Formula Weight 260.0128
Chemical Name or Material Phorate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.