missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phorate-d10 (O,O-diethyl-d10), CDN
Click to view available options
Quantity:
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | PhorateD10 |
| CAS | 1219805-45-4 |
| Molecular Formula | C7 2H10 H7 O2 P S3 |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C7H17O2PS3/c1-4-8-10(11,9-5-2)13-7-12-6-3/h4-7H2,1-3H3/i1D3,2D3,4D2,5D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])OP(=S)(OC([2 H])([2 H])C([2 H])([2 H])[2 H])SCSCC |
| IUPAC Name | ethylsulfanylmethylsulfanyl-bis(1,1,2,2,2-pentadeuterioethoxy)-sulfanylidene-λ^{5}-phosphane |
| Molecular Weight (g/mol) | 270.44 |
| Formula Weight | 270.0756 g/mol |
| Percent Purity | 99 atom % D, min 95% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction