missing translation for 'onlineSavingsMsg'
Learn More

Phorate Sulfoxide, TRC

Product Code. 30134842
Change view
Click to view available options
Quantity:
100 mg
250 mg
2.5 g
Unit Size:
100mg
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134842 2.5 g 2.5g
30134827 100 mg 100mg
30007123 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134842 Supplier Toronto Research Chemicals Supplier No. TRCP3534902.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2588-03-6
Molecular Formula C7 H17 O3 P S3
Molecular Weight (g/mol) 276.38
Synonym Phosphorodithioic acid, O,O-diethyl (ethylsulfinyl)methyl ester (6CI), Methanethiol, (ethylsulfinyl)-, S-ester with O,O-diethyl phosphorodithioate (8CI), Phorate O-sulfoxide, Phorate sulfoxide, Thimet sulfoxide
IUPAC Name diethoxy-(ethylsulfinylmethylsulfanyl)-sulfanylidene-λ^{5}-phosphane
SMILES CCOP(=S)(OCC)SCS(=O)CC

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C7H17O3PS3/c1-4-9-11(12,10-5-2)13-7-14(8)6-3/h4-7H2,1-3H3
Formula Weight 276.0077
Chemical Name or Material Phorate-sulfoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.