missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Phorbol 12,13-dibutyrate

Product Code. 18015942
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18015942 1 mg 1mg
18005632 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18015942 Supplier R&D Systems™ Supplier No. 4153/1
  • ONLINE EXCLUSIVE PRICE
    137.09 €
    118.65 € / 1mg
    Save 18.44 €
    13% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Protein kinase C activator

Phorbol 12,13-dibutyrate is a protein kinase C activator. Induces contraction of vascular smooth muscle and inhibits MLC phosphatase (MLCP) in vascular smooth muscle. Activity does not alter intracellular Ca2+ concentration. Also inhibits the activity of Na+,K+ ATPase in OK cells.

For more information about how Phorbol 12,13-dibutyrate may be used, see our protocol:

Generation of β cells from hPSCs
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37558-16-0
Molecular Formula C28H40O8

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 504.61
Purity >98%
Molecular Weight (g/mol) 504.62
Chemical Name or Material Phorbol 12, 13-dibutyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.