missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Phorbol 12-myristate 13-acetate

Product Code. 18752704
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18752704 1 mg 1mg
18772554 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18752704 Supplier R&D Systems™ Supplier No. 1201/1
  • ONLINE EXCLUSIVE PRICE
    151.42 €
    135.45 € / 1mg
    Save 15.97 €
    10% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Protein kinase C activator

Phorbol 12-myristate 13-acetate is an extensively used phorbol ester high affinity activator of protein kinase C (Ki = 2.6 nM as measured by displacement of [3H]phorbol 12,13-dibutyrate binding in rat cortex synaptosomal membranes). It binds to the C1a and C1b domains of PKC inducing membrane translocation of PKC. Phorbol 12-myristate 13-acetate activates the PKC isoforms PKCα, -β, -γ, -δ, -ε, -η, and -θ but not PKCζ or -ι/λ. Phorbol 12-myristate 13-acetate is a tumor promoter. Phorbol 12-myristate 13-acetate can also induce differentiation of THP-1 monocytes to a macrophage phenotype in vitro.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 16561-29-8
Molecular Formula C36H56O8

Specifica

Quantity 1 mg
Product Line Tocris
Formula Weight 616.83
Purity >99%
Molecular Weight (g/mol) 616.84
Chemical Name or Material Phorbol 12-myristate 13-acetate
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.