missing translation for 'onlineSavingsMsg'
Learn More

Phorbol 12-myristate 13-acetate, MedChemExpress

Product Code. p-200098853
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
Product Code. Quantity unitSize
16451749 1 mg 1mg
16423616 25 mg 25mg
16410088 5 mg 5mg
16417618 10 mg 10mg
4 options
This item is not returnable. View return policy

Product Code. 16451749

Brand: Medchem Express HY18739

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Phorbol 12-myristate 13-acetate (PMA), a phorbol ester, is a dual SphK and protein kinase C (PKC) activator. Phorbol 12-myristate 13-acetate is a NF-κB activator. Phorbol 12-myristate 13-acetate induces differentiation in THP-1 cells.

Phorbol 12-myristate 13-acetate (PMA), a phorbol ester, is a dual SphK and protein kinase C (PKC) activator. Phorbol 12-myristate 13-acetate is a NF-κB activator. Phorbol 12-myristate 13-acetate induces differentiation in THP-1 cells.

Specifications

Chemical Name or Material Phorbol 12-myristate 13-acetate
CAS 16561-29-8
Color White
Purity Grade Notes Research
Molecular Formula C36H56O8
Quantity 1 mg
Synonym PMA TPA Phorbol myristate acetate
Solubility Information DMSO : 100 mg/mL (162.12 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (162.12 mM; Need ultrasonic)
SMILES CCCCCCCCCCCCCC(O[C@H]([C@H]1C)[C@]2(OC(C)=O)[C@@]([C@@](C=C(CO)C[C@]34O)([H])[C@@]1(O)[C@]4([H])C=C(C)C3=O)([H])C2(C)C)=O
Molecular Weight (g/mol) 616.83
Formula Weight 616.83
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.