missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phosphate, 0.2M buffer soln., pH 7.2

Product Code. 15485189
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
This item is not returnable. View return policy

Product Code. 15485189

Brand: Thermo Scientific Chemicals J62899.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7558-80-7119.976 g/mol

Sodium phosphate monobasic is a reagent with very high buffering capacity widely used in molecular biology, biochemistry and chromatography. Useful in conjunction with Sodium Phosphate, Dibasic in the preparation of biological buffers. Used in many applications including the purification of antibodies. It is also used in HPLC. Phosphates are often used in foods and in water treatment. It is added in animal feed, toothpaste, and evaporated milk. It is used as a thickening agent and emulsifier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 7558-80-7
Molecular Weight (g/mol) 119.976

Specifikationer

Concentration 0.2M buffer soln.
Physical Form Liquid
Quantity 500 mL
Packaging Plastic Bottle

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.