missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phosphate, 0.5M buffer soln., pH 7.5

Product Code. 15415609
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15415609 500 mL 500mL
15405609 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15415609 Supplier Thermo Scientific Chemicals Supplier No. J63349.AP
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7558-80-7119.976 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-80-7
Molecular Weight (g/mol) 119.976

Specifications

Concentration 0.5M buffer soln.
Physical Form Liquid
Quantity 500 mL
Packaging Plastic Bottle

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.