missing translation for 'onlineSavingsMsg'
Learn More

Phosphinic Acid, puriss. p.a., 50% (T), Honeywell™

Product Code. 15600530 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Poly Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15600530 100 mL Poly Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15600530 Supplier Honeywell Research Chemicals Supplier No. 00651100ML
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Appearance Liquid
pH 1 (at 20°C)
Color Colorless
CAS 6303-21-5
Composition 0.5
Melting Point <-25°C
Boiling Point 108°C
Density 1.206g/mL at 20°C
Packaging Poly Bottle
Vapor Pressure 30 hPa at 20°C
Quantity 100 mL
Molecular Formula H3O2P
Linear Formula H3PO2
MDL Number MFCD02183592
UN Number UN3264
Synonym hypophosphite ion, hypophosphorus acid, phosphinic acid, ion 1-, hydroxyphosphanone, oxo-, e5-phosphanol, hydroxidooxidophosphorus, perfluoroalkyl c6-c12 phosphonic acid, diperfluoroalkyl c6-c12 phosphinic acid, hydroxy-, e5-phosphanone, perfluoro-c6-c12 ethyl hydrogen phosphate
InChI Key ACVYVLVWPXVTIT-UHFFFAOYSA-N
SMILES O[PH2]=O
IUPAC Name phosphenous acid
Molecular Weight (g/mol) 66.00
PubChem CID 22497
ChEBI CHEBI:36363
Formula Weight 66g/mol
Percent Purity 50% (T)
Grade puriss. p.a.
Chemical Name or Material Phosphinic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.