missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Phosphodiesterase 4 Inhibitor
99.70 € - 329.00 €
Specifications
Description | Pale Yellow |
---|---|
Content And Storage | 2°C to 8°C |
For Use With (Application) | Pyrazole compound with a high-affinity active site binding inhibitor of phosphodiesterases IVB and IVD |
CAS | 346440-86-6 |
Molecular Formula | C14H15N3O4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15426399
|
Thermo Scientific™
J64453.MB |
25 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15416399
|
Thermo Scientific™
J64453.#0 |
5 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Pyrazole compound with a high-affinity active site binding inhibitor of phosphodiesterases IVB and IVD.Specifications
Pale Yellow | |
Pyrazole compound with a high-affinity active site binding inhibitor of phosphodiesterases IVB and IVD | |
C14H15N3O4 | |
Ethyl 3,5-dimethyl-1-(3-nitrophenyl)-1H-pyrazole-4-carboxylate | |
Signal Transduction Reagent |
2°C to 8°C | |
346440-86-6 | |
289.29g/mol | |
Light Sensitive | |
Solid |