missing translation for 'onlineSavingsMsg'
Learn More

Phosphoenolpyruvic acid monopotassium salt, 99%

Product Code. 11454257
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11454257

Brand: Thermo Scientific Alfa Aesar B20358.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4265-07-0 | C3H4KO6P | 206.131 g/mol

Phosphoenolpyruvic acid monopotassium salt is used in glycolysis and gluconeogeneis. It is used for regeneration of ATP from ADP in enzyme-catalyzed syntheses. It is metabolized by an enzyme namely pyruvate kinase to yield pyruvate. It acts as protection against cell injury induced by oxidative stress and against dysfunction in carbohydrate metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphoenolpyruvic acid monopotassium salt is used in glycolysis and gluconeogeneis. It is used for regeneration of ATP from ADP in enzyme-catalyzed syntheses. It is metabolized by an enzyme namely pyruvate kinase to yield pyruvate. It acts as protection against cell injury induced by oxidative stress and against dysfunction in carbohydrate metabolism.

Solubility
Soluble in water. Slightly soluble in methanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4265-07-0
Molecular Formula C3H4KO6P
Molecular Weight (g/mol) 206.131
MDL Number MFCD00044476
InChI Key SOSDSEAIODNVPX-UHFFFAOYSA-M
Synonym potassium 1-carboxyvinyl hydrogenphosphate, potassium hydrogen 2-phosphonatooxy acrylate, 2-propenoic acid, 2-phosphonooxy-, monopotassium salt, potassium 1-carboxyvinyl hydrogen phosphate, phospho enol pyruvic acid monopotassium salt, phosphoenolpyruvic acid monopotassium salt, 2-propenoic acid, 2-phosphonooxy-, potassium salt 1:1, potassium 2-hydrogen phosphonooxy prop-2-enoic acid, phosphoenolpyruvate potassium salt, potassium 2-hydrogen phosphonatooxy prop-2-enoic acid
PubChem CID 23678879
IUPAC Name potassium;1-carboxyethenyl hydrogen phosphate
SMILES C=C(C(=O)O)OP(=O)(O)[O-].[K+]

Specifications

Melting Point ∼170°C (decomposition)
Assay Percent Range 0.99
Quantity 25 g
Beilstein 4603446
Sensitivity Hygroscopic
Solubility Information Soluble in water. Slightly soluble in methanol.
Formula Weight 206.14
Percent Purity 99%
Chemical Name or Material Phosphoenolpyruvic acid monopotassium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.