Learn More
Abcam Phosphoenolpyruvic acid (PEP) Assay Kit
Product Code. 32120626 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
Product Code. Quantity Unit Size
32120626 100 Tests 100 tests
1 options
Product Code. 32120626 Supplier Abcam Supplier No. ab204713100Test
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Measure Phosphoenolpyruvate activity in Cell/Tissue Extracts, Biofluids with Phosphoenolpyruvic acid (PEP) Assay Kit AB204713. Sensitivity = 1 μM. Cited in 5 publications. Use with Microplate reader.
Measure Phosphoenolpyruvate activity in Cell/Tissue Extracts, Biofluids with Phosphoenolpyruvic acid (PEP) Assay Kit AB204713. Sensitivity = 1 μM. Cited in 5 publications. Use with Microplate reader.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 100 Tests