missing translation for 'onlineSavingsMsg'
Learn More

Phosphomolybdic Acid Hydrate, TRC

Product Code. 30138371
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138371 5 g 5g
30135764 10 g 10g
30010294 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138371 Supplier Toronto Research Chemicals Supplier No. TRCP3591905G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51429-74-4
Molecular Formula H3[P(Mo3O10)4]
Molecular Weight (g/mol) 1825.25
Synonym Molybdic Phosphoric Anhydride Hydrate, Molybdophosphoric Acid (H3Mo12PO40) Hyrate, Molybdophosphoric Acid (H3PMo12O40), Hydrate, Molybdophosphoric Acid (H6P2Mo24O80), Hydrate, PMA, Molybdophosphoric Acid, Nonacosahydrate, Molybdate(3-), Tetracosa-μ-oxododecaoxo[μ12-[phosphato(3-)-kO:kO:kO:kO':kO':kO':kO' ':kO' ':kO' ':kO' ' ':kO' ' ':kO' ' ']]dodeca-, trihydrogen, Hydrate;
SMILES O.O[Mo]1234O[Mo]567(O)O[Mo]89(=O)(O1)O[Mo]%10%11%12(=O)O[Mo]%13%14%15(=O)O[Mo]%16%17%18(O)O[Mo]%19%20(=O)(O2)O[Mo](=O)(O%16)(O%10)(O8)O%17%19=P%21(O359)O%11%13[Mo](=O)(O6)(O%12)(O%14)O[Mo]%22%23(=O)(O7)O[Mo]%24(=O)(O%18)(O[Mo](=O)(O4)(O%22)(O%20)O%21%23%24)O%15

Specifications

Quantity 5 g
InChI Formula InChI=1S/12Mo.O4P.4H2O.33O/c;;;;;;;;;;;;1-5(2,3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h;;;;;;;;;;;;;4*1H2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/q;;;;;;;;;3*+1;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/p-3
Formula Weight 1866.6693
Chemical Name or Material Phosphomolybdic Acid Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.